Lithium 2-(6-(1H-imidazol-1-yl)pyridazine-3-carboxamido)-4,5-difluorobenzoate 98%

Supplier: Ambeed
Warning

Synonyms: SR-717

A1340104-100MG A1340104-250MG A1340104-50MG
77566-616EA 337.84 USD
77566-616 77566-618 77621-616
Lithium 2-(6-(1H-imidazol-1-yl)pyridazine-3-carboxamido)-4,5-difluorobenzoate 98%
Lithium 2-(6-(1H-imidazol-1-yl)pyridazine-3-carboxamido)-4,5-difluorobenzoate

Precaución: For research use only.

Formula: C₁₅H₈F₂LiN₅O₃
MW: 351.19 g/mol
CAS Number: 2375421-09-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR