(S)-N-Boc-2-(2-Hydroxyethyl)morpholine 97%

Supplier: Ambeed
Warning

A133870-1G A133870-250MG
77566-610EA 198.79 USD
77566-610 77666-928
(S)-N-Boc-2-(2-Hydroxyethyl)morpholine 97%
(S)-N-Boc-2-(2-Hydroxyethyl)morpholine

Precaución: For research use only.

Formula: C₁₁H₂₁NO₄
MW: 231.28 g/mol
MDL Number: MFCD13194712
CAS Number: 1257856-15-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR