DMG-PEG2000

Supplier: Ambeed
Warning

Synonyms: PEG 2000, DMG-PEG PEG 2000, 1,2-Dimyristoyl-rac-glycero-3-methoxypolyethylene glycol-2, Poly(oxy-1,2-ethanediyl), α-[2,3-bis[(1-oxotetradecyl)oxy]propyl]-ω-methoxy-

A1326597-1G A1326597-250MG A1326597-100MG
77566-576EA 373.89 USD
77566-576 77566-578 77566-574
DMG-PEG2000
DMG-PEG2000

Precaución: For research use only.

Formula: (C₂H₄O)nC₃₂H₆₂O₅
CAS Number: 160743-62-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR