tert-Butyl 3-(prop-2-yn-1-yl)azetidine-1-carboxylate 98%

Supplier: Ambeed
Warning

A1326116-250MG A1326116-1G
77566-570EA 131.84 USD
77566-570 77621-510
tert-Butyl 3-(prop-2-yn-1-yl)azetidine-1-carboxylate 98%
tert-Butyl 3-(prop-2-yn-1-yl)azetidine-1-carboxylate

Precaución: For research use only.

Formula: C₁₁H₁₇NO₂
MW: 195.258 g/mol
MDL Number: MFCD32173212
CAS Number: 1463502-41-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR