(E)-Prop-1-en-1-ylboronic acid 98%

Supplier: Ambeed
Warning

A130800-1G A130800-100MG A130800-250MG
77566-540EA 205 USD
77566-540 77883-051 77883-052
(E)-Prop-1-en-1-ylboronic acid 98%
(E)-Prop-1-en-1-ylboronic acid

Precaución: For research use only.

Formula: C₃H₇BO₂
MDL Number: MFCD02179501
CAS Number: 7547-97-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR