trans-4-Methylcyclohexylamine 98%, Ambeed.Inc

Supplier: Ambeed
Danger

A130421-5G A130421-1G A130421-25G
77566-534EA 18 USD
77566-534 77683-159 77683-160
trans-4-Methylcyclohexylamine 98%, Ambeed.Inc
trans-4-Methylcyclohexylamine

Precaución: For research use only.

Formula: C₇H₁₅N
MDL Number: MFCD06411232
CAS Number: 2523-55-9
UN: 2733
ADR: 3,8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR