Salicyl alcohol 99%

Supplier: Ambeed
Warning

A130074-100G A130074-25G A130074-5G
77566-526EA 38 USD
77566-526 77566-528 77611-150
Salicyl alcohol 99%
Salicyl alcohol

Precaución: For research use only.

Formula: C₇H₈O₂
MDL Number: MFCD00004617
CAS Number: 90-01-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR