1-(Methoxymethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole 95%

Supplier: Ambeed
Warning

A129611-1G A129611-250MG
77566-516EA 111.24 USD
77566-516 77745-935
1-(Methoxymethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole 95%
1-(Methoxymethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

Precaución: For research use only.

Formula: C₁₁H₁₉BN₂O₃
MW: 238.09 g/mol
MDL Number: MFCD28128543
CAS Number: 1301198-65-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR