Imidazo[1,2-a]pyrimidine-3-carboxylic acid 97%

Supplier: Ambeed
Warning

A129274-1G A129274-250MG A129274-100MG
77566-512EA 103 USD
77566-512 77592-496 77889-887
Imidazo[1,2-a]pyrimidine-3-carboxylic acid 97%
Imidazo[1,2-a]pyrimidine-3-carboxylic acid

Precaución: For research use only.

Formula: C₇H₅N₃O₂
MDL Number: MFCD09994697
CAS Number: 64951-11-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR