Neohesperidin dihydrochalcone 95%

Supplier: Ambeed
Warning

Synonyms: NHDC, NCI-c60764, Neohesperidin DC, 1-(4-(((2S,3R,4S,5S,6R)-4,5-Dihydroxy-6-(hydroxymethyl)-3-(((2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2-yl)oxy)-2,6-dihydroxyphenyl)-3-(3-hydroxy-4-methoxyphenyl)propan-1-one

A129040-5G A129040-100G A129040-25G
77566-508EA 17.51 USD
77566-508 77666-848 77666-850
Neohesperidin dihydrochalcone 95%
Neohesperidin dihydrochalcone

Precaución: For research use only.

Formula: C₂₈H₃₆O₁₅
MW: 612.58 g/mol
MDL Number: MFCD00017711
CAS Number: 20702-77-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR