3,5-Dimethoxybenzoic acid 98%

Supplier: Ambeed
Warning

A128578-10G A128578-100G A128578-500G
77566-498EA 6.18 USD
77566-498 77599-874 77670-824
3,5-Dimethoxybenzoic acid 98%
3,5-Dimethoxybenzoic acid

Precaución: For research use only.

Formula: C₉H₁₀O₄
MW: 182.18 g/mol
MDL Number: MFCD00002502
CAS Number: 1132-21-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR