4-(Azetidin-3-yl)morpholine hydrochloride 95%

Supplier: Ambeed
Warning

A128136-5G A128136-1G A128136-250MG
77566-490EA 267.8 USD
77566-490 77592-488 77683-136
4-(Azetidin-3-yl)morpholine hydrochloride 95%
4-(Azetidin-3-yl)morpholine hydrochloride

Precaución: For research use only.

Formula: C₇H₁₄N₂O·ClH
MW: 178.66 g/mol
MDL Number: MFCD11865371
CAS Number: 223381-71-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR