tert-Butyl 2-((2-(2,6-dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)oxy)acetate 98%

Supplier: Ambeed
Warning

A1273257-1G A1273257-5G
77566-472EA 193 USD
77566-472 77867-155
tert-Butyl 2-((2-(2,6-dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)oxy)acetate 98%
tert-Butyl 2-((2-(2,6-dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)oxy)acetate

Precaución: For research use only.

Formula: C₁₉H₂₀N₂O₇
MDL Number: MFCD32643096
CAS Number: 1950635-36-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR