Methyl 4-amino-2-ethylbenzoate 95%

Supplier: Ambeed
Warning

A126773-5G A126773-1G A126773-100MG
77566-458EA 645 USD
77566-458 77571-590 77747-909
Methyl 4-amino-2-ethylbenzoate 95%
Methyl 4-amino-2-ethylbenzoate

Precaución: For research use only.

Formula: C₁₀H₁₃NO₂
MDL Number: MFCD00229826
CAS Number: 1211589-24-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR