Tetrabutylammonium dibutyl phosphate 97%

Supplier: Ambeed
Warning

A1264123-1G A1264123-250MG
77566-444EA 208 USD
77566-444 77566-446
Tetrabutylammonium dibutyl phosphate 97%
Tetrabutylammonium dibutyl phosphate

Precaución: For research use only.

Formula: C₂₄H₅₄NO₄P
MW: 451.664 g/mol
CAS Number: 29536-33-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR