Bicyclo[1.1.1]pentan-1-ylboronic acid 98%

Supplier: Ambeed
Warning

A1263639-100MG A1263639-250MG
77566-440EA 697.31 USD
77566-440 77571-570
Bicyclo[1.1.1]pentan-1-ylboronic acid 98%
Bicyclo[1.1.1]pentan-1-ylboronic acid

Precaución: For research use only.

Formula: C₅H₉BO₂
MW: 111.935 g/mol
MDL Number: MFCD31705661
CAS Number: 2412765-14-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR