endo-BCN-PEG2-NHS ester 99+%

Supplier: Ambeed
Warning

A1255963-100MG A1255963-25MG A1255963-250MG
77566-422EA 384.19 USD
77566-422 77596-604 77621-308
endo-BCN-PEG2-NHS ester 99+%
endo-BCN-PEG2-NHS ester

Precaución: For research use only.

Formula: C₂₂H₃₀N₂O₈
MW: 450.482 g/mol
CAS Number: 2243565-12-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR