4,4,5,5-Tetramethyl-2-(1,2,2-triphenylvinyl)-1,3,2-dioxaborolane 98%

Supplier: Ambeed
Warning

A1250532-5G A1250532-1G
77566-408EA 268.83 USD
77566-408 77596-550
4,4,5,5-Tetramethyl-2-(1,2,2-triphenylvinyl)-1,3,2-dioxaborolane 98%
4,4,5,5-Tetramethyl-2-(1,2,2-triphenylvinyl)-1,3,2-dioxaborolane

Precaución: For research use only.

Formula: C₂₆H₂₇BO₂
MW: 382.3 g/mol
CAS Number: 219488-97-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR