4-Methoxy-2-methylpyrimidin-5-amine 98%

Supplier: Ambeed
Warning

A124763-1G A124763-250MG
77566-400EA 74 USD
77566-400 77881-002
4-Methoxy-2-methylpyrimidin-5-amine 98%
4-Methoxy-2-methylpyrimidin-5-amine

Precaución: For research use only.

Formula: C₆H₉N₃O
MDL Number: MFCD09743865
CAS Number: 53135-45-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR