3-(4-Bromophenyl)piperidine-2,6-dione 98%

Supplier: Ambeed
Warning

A1246271-1G A1246271-5G A1246271-100MG A1246271-250MG A1246271-25G
77566-392EA 27 USD
77566-392 77571-478 77866-982 77866-983 77866-984
3-(4-Bromophenyl)piperidine-2,6-dione 98%
3-(4-Bromophenyl)piperidine-2,6-dione

Precaución: For research use only.

Formula: C₁₁H₁₀BrNO₂
MDL Number: MFCD19372740
CAS Number: 1267337-47-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR