(2R,3S)-3-Isopropyl-2-phenyl-3,4-dihydro-2H-pyrimido[2,1-b][1,3]benzothiazole 98%

Supplier: Ambeed
Warning

A1237009-100MG A1237009-250MG A1237009-1G
77566-364EA 135 USD
77566-364 77596-514 77621-200
(2R,3S)-3-Isopropyl-2-phenyl-3,4-dihydro-2H-pyrimido[2,1-b][1,3]benzothiazole 98%
(2R,3S)-3-Isopropyl-2-phenyl-3,4-dihydro-2H-pyrimido[2,1-b][1,3]benzothiazole

Precaución: For research use only.

Formula: C₁₉H₂₀N₂S
MW: 308.4405 g/mol
MDL Number: MFCD29037150
CAS Number: 1699751-03-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR