Tetraethyl-4-tert-butylcalix[4]arene-O,O',O'',O'''-tetraacetate 97%

Supplier: Ambeed
Warning

Synonyms: 4-tert-Butylcalix[4]arene-O,O',O'',O'''-tetraacetic Acid Tetraethyl Ester, Tetraethyl 2,2',2'',2'''-((15,35,55,75-tetra-tert-butyl-1,3,5,7(1,3)-tetrabenzenacyclooctaphane-12,32,52,72-tetrayl)tetrakis(oxy))tetraacetate

A123445-1G A123445-250MG
77566-358EA 23.69 USD
77566-358 77624-030
Tetraethyl-4-tert-butylcalix[4]arene-O,O',O'',O'''-tetraacetate 97%
Tetraethyl-4-tert-butylcalix[4]arene-O,O',O'',O'''-tetraacetate

Precaución: For research use only.

Formula: C₆₀H₈₀O₁₂
MW: 993.29 g/mol
MDL Number: MFCD00145373
CAS Number: 97600-39-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR