3-(Triethoxysilyl)propyl 2-Bromo-2-methylpropanoate 95%

Supplier: Ambeed
Danger

Synonyms: 3-(Triethoxysilyl)propyl 2-bromo-2-methylpropanoate, 2-Bromo-2-methylpropanoic acid 3-(Triethoxysilyl)propyl ester

A1232571-5G A1232571-1G
77566-350EA 122.57 USD
77566-350 77571-418
3-(Triethoxysilyl)propyl 2-Bromo-2-methylpropanoate 95%
3-(Triethoxysilyl)propyl 2-Bromo-2-methylpropanoate

Precaución: For research use only.

Formula: C₁₃H₂₇BrO₅Si
MW: 371.34 g/mol
MDL Number: MFCD05865083
CAS Number: 880339-31-1
UN: 1760
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR