tert-Butyl ((S)-1-(((S)-1-((4-(hydroxymethyl)phenyl)amino)-1-oxopropan-2-yl)amino)-3-methyl-1-oxobutan-2-yl)carbamate 97%

Supplier: Ambeed
Warning

Synonyms: Boc-Val-Ala-PAB

A1227481-1G A1227481-100MG A1227481-250MG
77566-334EA 451.14 USD
77566-334 77596-462 77621-158
tert-Butyl ((S)-1-(((S)-1-((4-(hydroxymethyl)phenyl)amino)-1-oxopropan-2-yl)amino)-3-methyl-1-oxobutan-2-yl)carbamate 97%
tert-Butyl ((S)-1-(((S)-1-((4-(hydroxymethyl)phenyl)amino)-1-oxopropan-2-yl)amino)-3-methyl-1-oxobutan-2-yl)carbamate

Precaución: For research use only.

Formula: C₂₀H₃₁N₃O₅
MW: 393.48 g/mol
MDL Number: MFCD28557280
CAS Number: 1884577-99-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR