Topotecan hydrochloride 98%

Supplier: Ambeed
Danger

A122669-1G A122669-100MG A122669-10G A122669-10MG A122669-5G A122669-250MG
77566-322EA 130 USD
77566-322 77599-772 77599-774 77599-776 77599-778 77666-760
Topotecan hydrochloride 98%
Topotecan hydrochloride

Precaución: For research use only.

Formula: C₂₃H₂₃N₃O₅.HCl
MDL Number: MFCD00866235
CAS Number: 119413-54-6
UN: 1544
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR