(2S,4R)-1-((S)-2-(1-Fluorocyclopropanecarboxamido)-3,3-dimethylbutanoyl)-4-hydroxy-N-(2-hydroxy-4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide 97%

Supplier: Ambeed
Warning

Synonyms: VH032-cyclopropane-F

A1220565-1G A1220565-250MG A1220565-25MG A1220565-50MG A1220565-100MG
77566-292EA 1856.06 USD
77566-292 77566-294 77571-340 77571-342 77621-102
(2S,4R)-1-((S)-2-(1-Fluorocyclopropanecarboxamido)-3,3-dimethylbutanoyl)-4-hydroxy-N-(2-hydroxy-4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide 97%
(2S,4R)-1-((S)-2-(1-Fluorocyclopropanecarboxamido)-3,3-dimethylbutanoyl)-4-hydroxy-N-(2-hydroxy-4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide

Precaución: For research use only.

Formula: C₂₆H₃₃FN₄O₅S
MW: 532.627 g/mol
MDL Number: MFCD32201089
CAS Number: 2306193-99-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR