(4S,4'S)-4,4',5,5'-Tetrahydro-4,4'-diphenyl-2,2'-bioxazole 97%
Supplier: Ambeed
|
Warning
|
A1219906-100MG
A1219906-250MG
77566-286EA
29
USD
77566-286
77566-288
(4S,4'S)-4,4',5,5'-Tetrahydro-4,4'-diphenyl-2,2'-bioxazole 97%
(4S,4'S)-4,4',5,5'-Tetrahydro-4,4'-diphenyl-2,2'-bioxazole
Precaución: For research use only.
Formula:
C₁₈H₁₆N₂O₂ MW: 292.33 g/mol |
CAS Number:
135532-33-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...