Methyl 2-cyanoacrylate ≥98% stabilized

Supplier: Ambeed
Warning


Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

A1219011-1G A1219011-5G A1219011-250MG
77566-272EA 62.83 USD
77566-272 77566-274 77571-302
Methyl 2-cyanoacrylate ≥98% stabilized
Methyl 2-cyanoacrylate

Precaución: For research use only.

Formula: C₅H₅NO₂
MW: 111.1 g/mol
MDL Number: MFCD00079082
CAS Number: 137-05-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR