3-(((1S,2S,3R)-2,3-Difluoro-1-hydroxy-7-(methylsulfonyl)-2,3-dihydro-1H-inden-4-yl)oxy)-5-fluorobenzonitrile 98%

Supplier: Ambeed
Warning

A1216748-25MG A1216748-100MG A1216748-50MG
77566-254EA 116 USD
77566-254 77571-292 77866-718
3-(((1S,2S,3R)-2,3-Difluoro-1-hydroxy-7-(methylsulfonyl)-2,3-dihydro-1H-inden-4-yl)oxy)-5-fluorobenzonitrile 98%
3-(((1S,2S,3R)-2,3-Difluoro-1-hydroxy-7-(methylsulfonyl)-2,3-dihydro-1H-inden-4-yl)oxy)-5-fluorobenzonitrile

Precaución: For research use only.

Formula: C₁₇H₁₂F₃NO₄S
MDL Number: MFCD32197212
CAS Number: 1672668-24-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR