Manganese(III) oxide 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Manganese oxide (Mn2O3), Dimanganese trioxide
A1215141-100G
A1215141-25G
77566-252EA
10.3
USD
77566-252
77620-934
Manganese(III) oxide 98%
Manganese(III) oxide
Precaución: For research use only.
Formula:
Mn₂O₃ MW: 157.87 g/mol |
MDL Number:
MFCD00016217 CAS Number: 1317-34-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...