4-Hydroxycyclohexyl methacrylate stabilised

Supplier: Ambeed
Warning


Stabilization: Stabilised with TBC

A1213659-25G A1213659-5G
77566-248EA 282 USD
77566-248 77866-694
4-Hydroxycyclohexyl methacrylate stabilised
4-Hydroxycyclohexyl methacrylate

Precaución: For research use only.

Formula: C₁₀H₁₆O₃
MW: 184.23 g/mol
CAS Number: 203245-10-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR