5'-O-DMT-2'-O-TBDMS-N-Bz-Adenosine 95%

Supplier: Ambeed
Warning

A120635-5G A120635-25G A120635-1G
77566-238EA 133.9 USD
77566-238 77611-024 77670-712
5'-O-DMT-2'-O-TBDMS-N-Bz-Adenosine 95%
5'-O-DMT-2'-O-TBDMS-N-Bz-Adenosine

Precaución: For research use only.

Formula: C₄₄H₄₉N₅O₇Si
MW: 787.97 g/mol
MDL Number: MFCD00080294
CAS Number: 81265-93-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR