2-Hydroxy-n-octanoic acid 95%

Supplier: Ambeed
Warning

A1194709-5G A1194709-1G A1194709-25G A1194709-100G
77566-204EA 45 USD
77566-204 77571-160 77620-740 77866-510
2-Hydroxy-n-octanoic acid 95%
2-Hydroxy-n-octanoic acid

Precaución: For research use only.

Formula: C₈H₁₆O₃
MDL Number: MFCD00014410
CAS Number: 617-73-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR