(3-Ethynylphenyl)methanamine 96%

Supplier: Ambeed
Warning

A118431-250MG A118431-100MG
77566-168EA 273.98 USD
77566-168 77571-098
(3-Ethynylphenyl)methanamine 96%
(3-Ethynylphenyl)methanamine

Precaución: For research use only.

Formula: C₉H₉N
MW: 131.17 g/mol
MDL Number: MFCD10698132
CAS Number: 615288-78-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR