Di-μ-chlorotetrakis[3,5-difluoro-2-[5-trifluoromethyl-2-pyridinyl-kN)phenyl-kC]diiridium(III) 98%

Supplier: Ambeed
Warning

A1175857-250MG A1175857-1G A1175857-100MG
77566-146EA 344.02 USD
77566-146 77571-050 77620-478
Di-μ-chlorotetrakis[3,5-difluoro-2-[5-trifluoromethyl-2-pyridinyl-kN)phenyl-kC]diiridium(III) 98%
Di-μ-chlorotetrakis[3,5-difluoro-2-[5-trifluoromethyl-2-pyridinyl-kN)phenyl-kC]diiridium(III)

Precaución: For research use only.

Formula: C₄₈H₂₀Cl₂F₂₀Ir₂N₄
MW: 1488.0073 g/mol
MDL Number: MFCD29037184
CAS Number: 870987-64-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR