(2R,4R)-1-(5-Chloro-2-((3-cyanobenzyl)oxy)-4-((3-(2,3-dihydrobenzo[b][1,4]dioxin-6-yl)-2-methylbenzyl)oxy)benzyl)-4-hydroxypyrrolidine-2-carboxylic acid 99%

Supplier: Ambeed
Warning

Synonyms: BMS-1166

A1168529-100MG A1168529-25MG A1168529-50MG A1168529-250MG
77566-134EA 686 USD
77566-134 77596-014 77596-016 77620-404
(2R,4R)-1-(5-Chloro-2-((3-cyanobenzyl)oxy)-4-((3-(2,3-dihydrobenzo[b][1,4]dioxin-6-yl)-2-methylbenzyl)oxy)benzyl)-4-hydroxypyrrolidine-2-carboxylic acid 99%
(2R,4R)-1-(5-Chloro-2-((3-cyanobenzyl)oxy)-4-((3-(2,3-dihydrobenzo[b][1,4]dioxin-6-yl)-2-methylbenzyl)oxy)benzyl)-4-hydroxypyrrolidine-2-carboxylic acid

Precaución: For research use only.

Formula: C₃₆H₃₃ClN₂O₇
MDL Number: MFCD31746874
CAS Number: 1818314-88-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR