2,2-Dimethyl-4-oxo-3,7,10,13,16-pentaoxanonadecan-19-oic acid 95%

Supplier: Ambeed
Warning

Synonyms: Acid-PEG4-C2-Boc

A1163640-1G A1163640-100MG A1163640-250MG A1163640-50MG
77566-104EA 475 USD
77566-104 77595-968 77595-970 77595-972
2,2-Dimethyl-4-oxo-3,7,10,13,16-pentaoxanonadecan-19-oic acid 95%
2,2-Dimethyl-4-oxo-3,7,10,13,16-pentaoxanonadecan-19-oic acid

Precaución: For research use only.

Formula: C₁₆H₃₀O₈
MDL Number: MFCD29042335
CAS Number: 1835759-85-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR