tert-Butyl (7-hydroxyheptyl)carbamate 95%

Supplier: Ambeed
Warning

Synonyms: tert-butyl N-(7-hydroxyheptyl)carbamate

A1161057-1G A1161057-100MG A1161057-250MG
77566-100EA 126.69 USD
77566-100 77620-304 77620-306
tert-Butyl (7-hydroxyheptyl)carbamate 95%
tert-Butyl (7-hydroxyheptyl)carbamate

Precaución: For research use only.

Formula: C₁₂H₂₅NO₃
MW: 231.3318 g/mol
MDL Number: MFCD02094527
CAS Number: 173436-02-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR