Ir[dFFppy]2-(4,4′-dCF3bpy)PF6 98%

Supplier: Ambeed
Warning

A1158732-100MG A1158732-250MG A1158732-1G
77566-090EA 179 USD
77566-090 77566-092 77620-286
Ir[dFFppy]2-(4,4′-dCF3bpy)PF6 98%
Ir[dFFppy]2-(4,4′-dCF3bpy)PF6

Precaución: For research use only.

Formula: C₃₄H₁₆F₁₈IrN₄P
CAS Number: 2030437-92-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR