3,3-Bis(bromomethyl)-3,4-dihydro-2H-thieno[3,4-b][1,4]dioxepine 98%

Supplier: Ambeed
Danger

A1156759-5G A1156759-1G
77566-086EA 219.39 USD
77566-086 77620-264
3,3-Bis(bromomethyl)-3,4-dihydro-2H-thieno[3,4-b][1,4]dioxepine 98%
3,3-Bis(bromomethyl)-3,4-dihydro-2H-thieno[3,4-b][1,4]dioxepine

Precaución: For research use only.

Formula: C₉H₁₀Br₂O₂S
MW: 342.0475 g/mol
CAS Number: 701209-98-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR