N-((1H-Benzo[d]imidazol-2-yl)methyl)-1-(pyridin-2-yl)-N-(pyridin-2-ylmethyl)methanamine 97%

Supplier: Ambeed
Warning

A1150919-100MG A1150919-250MG
77566-054EA 174.07 USD
77566-054 77570-882
N-((1H-Benzo[d]imidazol-2-yl)methyl)-1-(pyridin-2-yl)-N-(pyridin-2-ylmethyl)methanamine 97%
N-((1H-Benzo[d]imidazol-2-yl)methyl)-1-(pyridin-2-yl)-N-(pyridin-2-ylmethyl)methanamine

Precaución: For research use only.

Formula: C₂₀H₁₉N₅
MW: 329.398 g/mol
MDL Number: MFCD31692663
CAS Number: 345349-15-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR