4-((6-Aminohexyl)oxy)-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione hydrochloride 99%

Supplier: Ambeed
Danger

Synonyms: Thalidomide-4-O-C6-NH2 hydrochloride

A1148745-1G A1148745-100MG
77566-040EA 687.01 USD
77566-040 77620-052
4-((6-Aminohexyl)oxy)-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione hydrochloride 99%
4-((6-Aminohexyl)oxy)-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione hydrochloride

Precaución: For research use only.

Formula: C₁₉H₂₄ClN₃O₅
MW: 409.8640 g/mol
MDL Number: MFCD31689259
CAS Number: 2245697-88-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR