4-((2-Aminoethyl)amino)-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: Pomalidomide-C2-NH2 HCl

A1148287-1G A1148287-250MG A1148287-100MG A1148287-50MG
77566-036EA 267 USD
77566-036 77570-854 77620-044 77620-046
4-((2-Aminoethyl)amino)-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione hydrochloride 95%
4-((2-Aminoethyl)amino)-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione hydrochloride

Precaución: For research use only.

Formula: C₁₅H₁₇ClN₄O₄
MW: 352.773 g/mol
MDL Number: MFCD31729301
CAS Number: 2305369-00-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR