Ethyl 6-bromo-2,2-dimethylhexanoate 97%

Supplier: Ambeed
Warning

A1147818-5G A1147818-1G
77566-032EA 48.41 USD
77566-032 77570-848
Ethyl 6-bromo-2,2-dimethylhexanoate 97%
Ethyl 6-bromo-2,2-dimethylhexanoate

Precaución: For research use only.

Formula: C₁₀H₁₉BrO₂
MW: 251.161 g/mol
MDL Number: MFCD22681601
CAS Number: 78712-62-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR