(((2R,3S,4R,5R)-5-(2-Amino-6-oxo-3,6-dihydro-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl) methyl) ((2R,3S,4R,5S,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl) dihydrogen diphosphate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Guanosine diphosphate fucose
A1147716-100MG
A1147716-250MG
A1147716-50MG
77566-030EA
313.12
USD
77566-030
77620-018
77620-020
(((2R,3S,4R,5R)-5-(2-Amino-6-oxo-3,6-dihydro-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl) methyl) ((2R,3S,4R,5S,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl) dihydrogen diphosphate 98%
(((2R,3S,4R,5R)-5-(2-Amino-6-oxo-3,6-dihydro-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl) methyl) ((2R,3S,4R,5S,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl) dihydrogen diphosphate
Precaución: For research use only.
Formula:
C₁₆H₂₅N₅O₁₅P₂ MW: 589.3417 g/mol |
MDL Number:
MFCD00870977 CAS Number: 15839-70-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...