4,4'-Dimethyldiphenyldisulfone 97%

Supplier: Ambeed
Warning

A1145182-250MG A1145182-1G
77566-020EA 69.01 USD
77566-020 77570-836
4,4'-Dimethyldiphenyldisulfone 97%
4,4'-Dimethyldiphenyldisulfone

Precaución: For research use only.

Formula: C₁₄H₁₄O₄S₂
MW: 310.389 g/mol
MDL Number: MFCD00159107
CAS Number: 10409-07-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR