2,2',2''-(1,4,7-Triazonane-1,4,7-triyl)triacetic acid 97%

Supplier: Ambeed
Warning

Synonyms: NOTA

A1143058-100MG A1143058-250MG A1143058-1G
77566-008EA 140.08 USD
77566-008 77570-792 77619-936
2,2',2''-(1,4,7-Triazonane-1,4,7-triyl)triacetic acid 97%
2,2',2''-(1,4,7-Triazonane-1,4,7-triyl)triacetic acid

Precaución: For research use only.

Formula: C₁₂H₂₁N₃O₆
MW: 303.312 g/mol
MDL Number: MFCD00137938
CAS Number: 56491-86-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR