(3R)-3-(Methoxymethyl)morpholine hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: (R)-3-(Methoxymethyl)morpholine hydrochloride

A114078-1G A114078-100MG A114078-250MG
77566-000EA 241.02 USD
77566-000 77610-914 77683-037
(3R)-3-(Methoxymethyl)morpholine hydrochloride 95%
(3R)-3-(Methoxymethyl)morpholine hydrochloride

Precaución: For research use only.

Formula: C₆H₁₃NO₂·ClH
MW: 167.634 g/mol
MDL Number: MFCD09608106
CAS Number: 218594-74-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR