(2S)-2-Acetamido-3,3-dimethylbutanoic acid 95%

Supplier: Ambeed
Warning

A1080104-1G A1080104-250MG A1080104-5G
77565-894EA 38 USD
77565-894 77570-462 77570-464
(2S)-2-Acetamido-3,3-dimethylbutanoic acid 95%
(2S)-2-Acetamido-3,3-dimethylbutanoic acid

Precaución: For research use only.

Formula: C₈H₁₅NO₃
MDL Number: MFCD08275827
CAS Number: 22146-59-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR