tert-Butyl 4-(prop-2-yn-1-yloxy)piperidine-1-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: tert-butyl 4-prop-2-ynoxypiperidine-1-carboxylate

A1076259-1G A1076259-250MG A1076259-100MG
77565-888EA 245 USD
77565-888 77570-454 77783-116
tert-Butyl 4-(prop-2-yn-1-yloxy)piperidine-1-carboxylate 95%
tert-Butyl 4-(prop-2-yn-1-yloxy)piperidine-1-carboxylate

Precaución: For research use only.

Formula: C₁₃H₂₁NO₃
MDL Number: MFCD16653309
CAS Number: 1219827-56-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR